missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LanthaScreen™ Tb-labeled Anti-Mouse Antibody Kit
LanthaScreen™ Tb-labeled Anti-Mouse Antibody Kit
Brand: Invitrogen™ PV3765
Code : 50
Additional Details : Weight : 0.05250kg
Description
To facilitate in-house assay development around multiple target classes and assay formats, Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents. Tb-labeled secondary antibodies bind to their corresponding species IgG with low nanomolar apparent Kd's, and show negligible cross reactivity with IgG's from other species. Each Tb-labeled anti-species antibody is provided with a corresponding fluorescein-labeled IgG as a positive control.In a kinase assay utilizing the Tb-labeled antibody generic reagent, anti-phosphopeptide primary antibody is not labeled and the detection step is performed by the addition of the Tb-labeled secondary antibody. This approach can be applied to other target classes and assay formats.
Specifications
Lanthanide Chelates, Terbium (Tb) | |
-5°C to -30°C | |
25 μg | |
Liquid | |
TR-FRET dilution buffer, LanthaScreen™ Tb-anti-mouse antibody, fluorescein-mouse IgG | |
LanthaScreen™ Tb-labeled Anti-Mouse Antibody Kit |
LanthaScreen™ Tb-labeled Anti-Mouse Secondary Antibody Kit, 25μg, Liquid, Mouse Species Reactivity, Mouse Host, IgG Detection Target, Terbium Conjugate, Fluorescent Detection Method, Water Soluble, 6 Months Shelf Life, Store in Freezer, Used for | |
Fluorescent | |
IgG | |
Liquid | |
Pharma and biopharma, secondary detection, target and lead identification and validation |
For Research Use Only. Not for use in diagnostic procedures.