missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LanthaScreen™ Tb-labeled Anti-rabbit Ab
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
Each
Beschreibung
A general reagent for use in assay development, this terbium labeled antibody will bind rabbit antibodies. This product is ideally suited for use during assay development when evaluating multiple primary rabbit antibodies or when you are unable to directly terbium label your primary antibody.
Spezifikation
Spezifikation
| Description | LanthaScreen™ Tb-labeled Anti-Rabbit Secondary Antibody, 25μg, Liquid, 1.03mg/mL Concentration, Rabbit Species Reactivity, Terbium Conjugate, Fluorescent Detection Method, 6 Months Shelf Life, Store in –20°C, Used for Pharma and Biophar |
| Content And Storage | Terbium-labeled anti-Rabbit antibody in HEPES buffered saline. Store at -20°C. |
| Detection Method | Fluorescence |
| Excitation/Emission | 340/490, 520 nm |
| Quantity | 25 μg |
| Reactivity | Rabbit |
| Readout | End Point |
| Shipping Condition | Dry Ice |
| Conjugate | Lanthanide Chelates, Terbium (Tb) |
| Format | Liquid |
| Mehr anzeigen |
For Research Use Only. Not for use in diagnostic procedures.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur