Learn More
Thermo Scientific™ LanthaScreen™ Tb-labeled Anti-Rabbit Antibody Kit
Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents to facilitate in-house assay development around multiple target classes and assay formats.
Brand: Thermo Scientific™ PV3775
Code : 50
Additional Details : Weight : 0.05250kg
Description
- Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents to facilitate in-house assay development around multiple target classes and assay formats
- Tb-labeled secondary antibodies bind to their corresponding species IgG with low nanomolar apparent Kd's, and show negligible cross reactivity with IgG's from other species
- Each Tb-labeled anti-species antibody is provided with a corresponding fluorescein-labeled IgG as a positive control
- In a kinase assay utilizing the Tb-labeled antibody generic reagent, anti-phosphopeptide primary antibody is not labeled
- The detection step is performed by the addition of the Tb-labeled secondary antibody
- This approach can be applied to other target classes and assay formats
Specifications
Lanthanide Chelates, Terbium (Tb) | |
Rabbit | |
1 mg |
Store in freezer (-5 to -30°C). | |
Dry Ice | |
LanthaScreen™ |
For Research Use Only. Not for use in diagnostic procedures.