missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LanthaScreen™ Tb-labeled Anti-Rabbit Antibody Kit
LanthaScreen™ Tb-labeled Anti-Rabbit Antibody Kit
Brand: Invitrogen™ PV3773
Code : 50
Additional Details : Weight : 0.05250kg
Description
To facilitate in-house assay development around multiple target classes and assay formats, Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents. Tb-labeled secondary antibodies bind to their corresponding species IgG with low nanomolar apparent Kd's, and show negligible cross reactivity with IgG's from other species. Each Tb-labeled anti-species antibody is provided with a corresponding fluorescein-labeled IgG as a positive control.In a kinase assay utilizing the Tb-labeled antibody generic reagent, anti-phosphopeptide primary antibody is not labeled and the detection step is performed by the addition of the Tb-labeled secondary antibody. This approach can be applied to other target classes and assay formats.
Specifications
Lanthanide Chelates, Terbium (Tb) | |
-5°C to -30°C | |
25 μg | |
Liquid | |
Pharma and biopharma, secondary detection, target and lead identification and validation, protein biology, cell analysis, drug discovery and development, target and lead identification and validation, kinase biology, kinase antibodies and immunoassays, bi |
LanthaScreen™ Tb-labeled Anti-Rabbit Secondary Antibody Kit, 25μg, Liquid, Rabbit Species Reactivity, IgG Isotype, Terbium Conjugate, Fluorescent Detection Method, Water Soluble, 6 Months Shelf Life, Store in Freezer, Used for Pharma and Biopharm | |
Fluorescent | |
Liquid | |
LanthaScreen™ Tb-anti-Rabbit antibody, Fluorescein-Rabbit IgG, TR-FRET dilution buffer | |
LanthaScreen™ Tb-labeled Anti-Rabbit Antibody Kit |
For Research Use Only. Not for use in diagnostic procedures.