Learn More
Invitrogen™ LanthaScreen™ Tb-peIF2a (pSer52) Antibody
The alpha subunit of Eukaryotic initiation factor 2 (eIF2alpha) plays a key regulatory role in protein synthesis.
Marke: Invitrogen™ PV4815
Code : 50
Weitere Details : Weight : 0.00900kg
Beschreibung
The alpha subunit of Eukaryotic initiation factor 2 (eIF2alpha) plays a key regulatory role in protein synthesis. In response to a cellular stress, phosphorylation of eIF2alpha at Ser52 occurs by the EIF2AK family of kinases including PERK (EIF2AK3) and PKR (EIF2AK2) to prohibit the initiation of protein synthesis.
For use in combination with the GFP-EIF2alpha physiological protein substrate (Cat. No. PV4809) to evaluate inhibitors in medium to high throughput screening applications.
Spezifikation
Serine/threonine kinase | |
0.47 mg/mL | |
25 μg | |
Pharma and Biopharma | |
1 Each | |
Liquid | |
Human | |
End Point | |
Ser/Thr Kinases |
Kinase Assay | |
Tb (Terbium)/Lanthanide Chelates | |
RUO | |
Primary | |
Terbium-labeled peIF2a (pThr46) antibody in HEPES buffered saline. Store at -20°C. | |
340/490, 520 nm | |
Dry Ice | |
Hu | |
LanthaScreen™ |
For Research Use Only. Not for use in diagnostic procedures.