missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Thiol Reactive Tb Chelate
LanthaScreen™ Thiol Reactive Tb Chelate
£261.00 - £10280.00
Specifications
Product Type | Thiol Reactive Tb Chelate |
---|---|
Content And Storage | LanthaScreen™ TR-FRET Labeling Reagents are provided lyophilized. Store at -20°C, desiccated, and protected from light. After reconstitution, terbium lanthanide chelates should be stored on ice and used within the same day. |
Shipping Condition | Dry Ice |
Product Line | LanthaScreen™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11543467
|
Thermo Scientific™
PV3580 |
10 μg |
£261.00
10µg |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
10082552
|
Thermo Scientific™
PV3578 |
1 mg |
£10280.00
1mg |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11533467
|
Thermo Scientific™
PV3579 |
100 μg |
£1396.00
100µg |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
Description
- As part of the LanthaScreen™ TR-FRET toolbox of assay reagents, LanthaScreen™ Thiol Reactive Tb Chelate labeling reagent is available for assay development
- The thiol-reactive maleimide group can be conjugated to virtually any peptide or protein that contains one or more accessible thiol moieties, such as the thiol group found on a cysteine residue
- Energy transfer from the terbium donor to a suitable acceptor such as fluorescein is readily detected by monitoring an increase in acceptor fluorescence intensity
- See the user guide for an application-based protocol for cysteine-containing peptides or proteins
Specifications
Thiol Reactive Tb Chelate | |
LanthaScreen™ TR-FRET Labeling Reagents are provided lyophilized. Store at -20°C, desiccated, and protected from light. After reconstitution, terbium lanthanide chelates should be stored on ice and used within the same day. | |
Dry Ice | |
LanthaScreen™ |
For Research Use Only. Not for use in diagnostic procedures.