Learn More
Invitrogen™ LanthaScreen™ TR-FRET Androgen Receptor Coactivator Assay Kit
LanthaScreen™ TR-FRET Androgen Receptor Coactivator Assay Kit
Brand: Invitrogen™ A15878
This item is not returnable.
View return policy
Description
The LanthaScreen™ TR-FRET Androgen Receptor Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential androgen receptor (AR) ligands as agonists of coactivator recruitment or antagonists of agonist-dependent coactivator recruitment. In both the agonist and antagonist modes, the homogeneous mix-and-read assay uses a rat AR ligand-binding domain (AR-LBD) tagged with hexahistidine and glutathione-S-transferase (GST) (also available separately), a terbium (Tb)-labeled anti-GST antibody, and a fluorescein-labeled coactivator peptide.Agonist Mode
When the LanthaScreen TR-FRET Androgen Receptor Coactivator Assay is run in agonist mode (to identify agonist compounds), AR-LBD is added to ligand test compounds, followed by the addition of a mixture of fluorescein-coactivator peptide and Tb-anti-GST antibody. After room temperature incubation, the TR-FRET 520:495 nm emission ratio is calculated and used to determine the EC50 from a dose response curve of the compound. This ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide (see figure).
Antagonist Mode
When the LanthaScreen TR-FRET Androgen Receptor Coactivator Assay is run in antagonist mode (to identify antagonist compounds), AR-LBD is added to ligand test compounds, followed by addition of a mixture of DHT agonist, fluorescein-coactivator peptide, and Tb-anti-GST antibody. The concentration of DHT agonist used in this mode is the EC80 concentration as determined by first running the assay in agonist mode. An example of data produced in the antagonist mode is shown in the figure below.
Specifications
1 tube AR Ligand Binding Domain Recombinant Protein (store at -68 to -85°C) 1 tube Peptide (store at -5 to -30°C) 2 bottles TR-FRET Buffer (store at -5 to -30°C) 1 tube Tb-Anti-GST Ab (store at -5 to -30°C) 1 tube of DTT (store at -5 to -30°C) |
|
Co-Factor Interaction Assay, Nuclear Receptor Assay | |
TR-FRET Androgen Receptor Coactivator Assay Kit |
LanthaScreen™ | |
AR | |
800 x 20 μL assays |
For Research Use Only. Not for use in diagnostic procedures.