Learn More
Cole-Parmer™ SH-200 Series See-Saw Rocking Shakers
Gently mix live cultures.
Brand:  Cole-Parmer™ 51901-06
Additional Details : Weight : 12.97270kg
 View more versions of this product
Product Code. 17721288
  • £1096.07 / Each
Estimated Shipment: 09-10-2024
to see stock.
Add to basket
Description
Description
  • Nonslip mat and platform lip hold vessels securely in place
  • Built-in timer and digital speed LED display provide reproducible results
  • Can be used in incubators
These see-saw rocking shakers provide a gentle rocking action at a 7° tilt angle, perfect for culture flasks and Petri dishes. Easily set and view the speed and the timer on the LED display. Control speed from 5 to 70 rpm. The microprocessor maintains speed accuracy even over long runs. Easily set digital timer from 1 to 999 minutes. When timer reaches zero, the shaker automatically stops and sounds an alert.Durable shakers are rated for continuous use. Use in incubators and environmental or cold chambers up to 104°F (40°C).
Specifications
Specifications
5 to 70 rpm
22 lb.
17.716 in.
6.5 in.
14 in.
15.35 in.
39 cm
Large See-Saw Rocker
Flat Platform, Nonslip Mat, and Power Cord
3 Year Warranty
10 kg
230 VAC
50 Hz
45 cm
16.5 cm
35.5 cm
14 in.
35.5 cm
LED
1 to 999 minutes