Learn More
Justrite™ Large Steel Drum Funnel With Self-Closing Cover
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ Large Steel Drum Funnel With Self-Closing Cover helps to prevent spills when transferring flammable solvents, thinners, oils, or paint wastes. This self-closing model meets NFPA1 and the International Fire Code. It shuts and locks automatically when a fusible link melts at 165°F (74°C) under fire conditions.
Marque: Justrite™ 08206
Code : 03
Informations supplémentaires : Weight : 5.50000kg
Description
- Justrite’s steel laboratory safety cans have all the benefits of the Type I cans
- Added features make these the choice of lab professionals for safely dispensing hazardous liquids into test tubes, small flasks and beakers
- Fill spouts have self-close, pressure-relief caps and stainless steel flame arresters
- These cans also feature a vacuum-relief valve within the lid for smooth liquid flow, and self-closing faucets for trouble-free dispensing
Spécification
Large Steel Drum Funnel | |
Flammables | |
32 in. flame arrester with 2 in. drum bung and self-closing cover |
Yellow | |
Steel (Galvanized) |