missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novacyt - Microgen Bioproducts Latex Agglutination Test: Staphylococcus aureus
Can be used to obtain rapid, presumptive-positive results from selective media for targeted identification/diagnosis.
£68.16 - £256.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Detectable Analytes | S. aureus |
Type | Latex Agglutination Test |
Test Time | 2 min. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15957112
|
Novacyt - Microgen Bioproducts
M433CE |
500 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15988012
|
Novacyt - Microgen Bioproducts
M43CE |
100 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17122199
|
Novacyt - Microgen Bioproducts
M43CE |
100 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Rapid (2 minute), simple, sensitive, and specific latex agglutination test for the identification and confirmation of infecting colonies.
- Can be used to obtain rapid, presumptive-positive results and provide an early indication of possible production process failure.
- Can be used to screen colonies before full biochemical identification and can replace traditional agglutinating antibody methods.
Specifications
2°C to 8°C | |
S. aureus | |
Latex Agglutination Test | |
2 min. |