missing translation for 'onlineSavingsMsg'
Learn More

Latex Count Drop Tubes

For use as temporary closures when glass blowing

Brand:  Fisherbrand LP/1

 View more versions of this product

Product Code. 12333569

  • £31.00 / Pack of 50
Estimated Shipment: 23-10-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Closed at one end for stirring rods
Specifications

Specifications

Bore size 3mm
Temporary closures when glass blowing
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers