Learn More
Fisherbrand™ Latex Dropper Teats
For easy dispensing of liquids.
£42.45 - £60.60
Products 5
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
12406180
View Documents
Fisherbrand™
DT/0/D001
1 mL
£46.04
Pack of 20
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
12416180
View Documents
Fisherbrand™
DT/1/DO01
1.5 mL
£42.45
Pack of 20
In Stock
to see stock.
Edge
Add to basket
12426180
View Documents
Fisherbrand™
DT/3/D001
2.5 mL
£44.01
Pack of 20
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
12436180
View Documents
Fisherbrand™
DT/5/D001
3.75 mL
£60.60
Pack of 20
In Stock
to see stock.
Edge
Add to basket
12446180
View Documents
Fisherbrand™
BVJ-382-000-B
1.5 to 5 mL (Assorted)
£49.50
Pack of 20
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Flexible rubber bulbs create suction and dispensing capacity when used with pipettes
  • Provide intuitive manual pipeting control
  • Available in different capacities
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Latex Dropper Teats