Learn More
Kartell™ Latex Dropping Bulb with Rim
Kartell™ Latex Dropping Bulb with Rim allows for easy dispensing of liquids and can be used with Ranvier Dropping Tubes and Pasteur Pipettes.
Brand: Kartell™ 0391400
This item is not returnable.
View return policy
Description
- Suitable for Ranvier dropping tubes
- Suitable for Pasteur pipettes
- Constructed of latex
Specifications
Latex | |
For Ranvier Dropping Tubes and Pasteur Pipettes | |
40 mm |
Dropping Bulb | |
With Rim |