Learn More
Novus Biologicals™ Latex and Europium Conjugated Antibody Purification Kit
Easy-to-use kit for the removal of interfering buffer components that can affect the conjugation reaction when using LATEX™ Conjugation Kits.
Brand: Novus Biologicals™ 1020-0040
Additional Details : Weight : 0.58200kg
Description
The kit utilizes a simple spin column to clean-up the antibody by buffer exchanging to remove the unwanted buffer components. The kit can also be used for concentrating your antibody by recovering the antibody in a smaller volume than the volume initially added to the Spin Cartridge.
Kit Contents:
- 1 vial 20x Reaction buffer A
- 1 vial 20x Reaction buffer B
- 4 spin cartridge / collecting tube assemblies
Specifications
4°C | |
4 Units | |
LATEX™ and Europium Conjugated |