missing translation for 'onlineSavingsMsg'
Learn More
Cole-Parmer™ North™ Powdered Latex Finger Cots Product Code.: 11736574

Cole-Parmer™ North™ Powdered Latex Finger Cots

Product Code. 11736574 Shop All Cole-Parmer Products
Medium
Click to view available options
Size:
Medium
This item is not returnable. View return policy

Product Code. 11736574

Brand: Cole-Parmer™ 8160542

Quantity Price
1 £39.64 / Pack of 1440
10 £36.35 / Pack of 1440

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.

View alternative products
This item is not returnable. View return policy

Use these finger cots when sensitivity and dexterity are needed. Cole-Parmer™ North™ Powdered Latex Finger Cots provide effective protection for delicate applications.

  • Meet ASTM D3772 for physical properties

Specifications

Certifications/Compliance ASTM D3772
Color Clear
Material Latex
Product Type Finger Cot
Size Medium
Thickness (English) 3 mil
For Use With (Application) Use when Sensitivity and Dexterity are needed
Quantity 1440Pack
Length (Metric) 68.85 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ North™ Powdered Latex Finger Cots >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.