missing translation for 'onlineSavingsMsg'
Learn More
Ansell™ Versatouch™ 78-103 Thermal Insulating Gloves Product Code.: 15785992
In Stock
to see stock.
Add to basket

Ansell™ Versatouch™ 78-103 Thermal Insulating Gloves

Product Code. 15785992 Shop All Ansell Products
9
Click to view available options
Size:
7
9

Product Code. 15785992

Brand: Ansell™ 78103090

In Stock
to see stock.
Add to basket
Add to basket

Offers protection from cold without sacrificing dexterity. Ansell Edmont™ Versatouch™ 78-103 Thermal Insulating Gloves are light-duty gloves with a seamless knit, thermal acrylic liner that provides comfort and warmth in cold or refrigerated work.

VersaTouch thermal insulating gloves are ideal to keep hands warm in refrigerated applications.
  • Uncoated liner for use alone or as an underglove
  • Seamless knit, thermal acrylic liner provides comfort and warmth
  • Acrylic liner offers protection from cold without sacrificing dexterity
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 388/511
Coating Material Uncoated
Color Blue
Exterior Finish Knitted
Grade 10 Guage
Latex Free Yes
Size 9
Sterility Non-sterile
For Use With (Application) Refrigerated/chilled areas, frozen products
Glove Cuff Style Knit
Liner Material Acrylic
Lining Uncoated
Packaging Type Bagged
Quantity 12 Pair
Length (Metric) 215 to 250 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell™ Versatouch™ 78-103 Thermal Insulating Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.