Learn More
Eurofins DiscoverX LATS2 Protein, Active
Active, N-Terminal GST-tagged, recombinant, human LATS2, amino acid residues 553-end and N-Terminal 6His-tagged, recombinant, human MOBKL1A, full length, for use in Kinase Assays.
Brand: Eurofins DiscoverX 14-987
Additional Details : Weight : 4.70710kg
Description
LATS2 is a serine/threonine protein kinase belonging to the LATS tumor suppressor family. The protein localizes to centrosomes during interphase, and early and late metaphase. It interacts with the centrosomal proteins aurora-A and ajuba and is required for accumulation of gamma-tubulin and spindle formation at the onset of mitosis. It also interacts with a negative regulator of p53 and may function in a positive feedback loop with p53 that responds to cytoskeleton damage. Additionally, it can function as a co-repressor of androgen-responsive gene expression.
GenBank NM_014572.2 / GenBank BC038112
Specifications
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
10 μg | |
Recombinant | |
Recombinant enzyme expressed in Sf21 insect cells | |
Combined ≥52% |
89 kDa (LATS2) and 30 kDa (MOBKL1A) | |
Human | |
N-terminal GST / N-terminal 6His | |
Truncated | |
LATS2, active |