missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lauryl chloride, 99%, Thermo Scientific Chemicals
£24.10 - £66.00
Chemical Identifiers
CAS | 112-52-7 |
---|---|
Molecular Formula | C12H25Cl |
Molecular Weight (g/mol) | 204.78 |
MDL Number | MFCD00013688 |
InChI Key | YAYNEUUHHLGGAH-UHFFFAOYSA-N |
Synonym | lauryl chloride, dodecyl chloride, n-dodecyl chloride, dodecane, 1-chloro, laurylchloride, unii-we8o8j0utk, 1-chloro-dodecane, dodecane, chloro, ccris 5810, we8o8j0utk |
PubChem CID | 8192 |
IUPAC Name | 1-chlorododecane |
SMILES | CCCCCCCCCCCCCl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10367990
|
Thermo Scientific Acros
257102500 |
250 mL | Glass bottle |
£24.10
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10056103
|
Thermo Scientific Acros
257100010 |
1 L | Glass bottle |
£66.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-52-7 | |
204.78 | |
YAYNEUUHHLGGAH-UHFFFAOYSA-N | |
8192 | |
CCCCCCCCCCCCCl |
C12H25Cl | |
MFCD00013688 | |
lauryl chloride, dodecyl chloride, n-dodecyl chloride, dodecane, 1-chloro, laurylchloride, unii-we8o8j0utk, 1-chloro-dodecane, dodecane, chloro, ccris 5810, we8o8j0utk | |
1-chlorododecane |
Specifications
112-52-7 | |
Colorless | |
260°C | |
Authentic | |
C12H25Cl | |
1.4420 to 1.444 | |
0.87 | |
Solubility in water: 0.0068g/L (20°C). Other solubilities: miscible with most common organic solvents,slightly miscible with alcohol,soluble in ethanol,benzene,ether,miscible with acetone | |
CCCCCCCCCCCCCl | |
204.78 | |
204.79 | |
Liquid |
-9.3°C | |
0.8700g/mL | |
130°C | |
98.5% min. (GC) | |
CH3(CH2)11Cl | |
MFCD00013688 | |
lauryl chloride, dodecyl chloride, n-dodecyl chloride, dodecane, 1-chloro, laurylchloride, unii-we8o8j0utk, 1-chloro-dodecane, dodecane, chloro, ccris 5810, we8o8j0utk | |
YAYNEUUHHLGGAH-UHFFFAOYSA-N | |
1-chlorododecane | |
8192 | |
99% | |
Lauryl chloride, 99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-981-5
missing translation for 'tsca' : TSCA
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Lauryl chloride
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
Precautionary Statement
- P273-Avoid release to the environment.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
RUO – Research Use Only