missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lauryl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | -23.0°C |
Color | Yellow |
Density | 0.8710g/mL |
Flash Point | 126°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Linear Formula | CH2=C(CH3)COO(CH2)11CH3 |
Refractive Index | 1.4440 to 1.4470 |
Quantity | 2.5 L |
Show More |
Safety and Handling
Product Identifier
- Lauryl methacrylate, stabilized
- Warning
- Specific target organ toxicity Category 3
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- MIXTURE LIST-Contains : Dodecyl methacrylate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction