missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lawesson's Reagent, 97%

Product Code. 11411898
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 11411898

Brand: Thermo Scientific Chemicals A14530.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19172-47-5 | C14H14O2P2S4 | 404.452 g/mol

Lawesson's reagent is a thiation agent used to convert carbonyl compounds into thiocarbonyls. It is also used to thionate enones, esters, lactones, amides, lactams and quinones. Further, it is used to prepare thiols from alcohols. It is associated with silver perchlorate and utilized as and oxophilic Lewis acid catalyst for Diels-Alder reaction of dienes with alfa, beta-unsaturated aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lawesson′s reagent is a thiation agent used to convert carbonyl compounds into thiocarbonyls. It is also used to thionate enones, esters, lactones, amides, lactams and quinones. Further, it is used to prepare thiols from alcohols. It is associated with silver perchlorate and utilized as and oxophilic Lewis acid catalyst for Diels-Alder reaction of dienes with alfa, beta-unsaturated aldehydes.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19172-47-5
Molecular Formula C14H14O2P2S4
Molecular Weight (g/mol) 404.452
MDL Number MFCD00005171
InChI Key CFHGBZLNZZVTAY-UHFFFAOYSA-N
Synonym lawesson's reagent, lawesson reagent, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane 2,4-disulfide, unii-a4125mq8rx, 1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-, 2,4-disulfide, 2,4-bis 4-methoxyphenyl-2,4-dithioxo-1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane-2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulphide, 4-methoxyphenylthiophosphoric cyclic di thioanhydride
PubChem CID 87949
IUPAC Name 2,4-bis(4-methoxyphenyl)-2,4-bis(sulfanylidene)-1,3,2$l^{5},4$l^{5}-dithiadiphosphetane
SMILES COC1=CC=C(C=C1)P2(=S)SP(=S)(S2)C3=CC=C(C=C3)OC

Specifications

Melting Point 221°C to 225°C
Quantity 100 g
UN Number UN3134
Beilstein 1024888
Sensitivity Moisture sensitive
Merck Index 14,5391
Solubility Information Insoluble in water.
Formula Weight 404.47
Percent Purity 97%
Chemical Name or Material Lawesson's Reagent
compliance-icons
Product Identifier
  • Lawesson's Reagent
Avvertenza
  • Danger
Categoria di pericolo
  • Substance which\, in contact with water\, emits flammable gases Category 2
Indicazioni di pericolo
  • H261-In contact with water releases flammable gases.
  • EUH029-Contact with water liberates toxic gas.
Consigli di prudenza
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Informazioni supplementari
  • MIXTURE LIST-Contains : 1,3,2,4-Dithiadiphosphetane, 2,4-bis(4-methoxyphenyl)-, 2,4-disulfide

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato