Learn More
Thermo Scientific Chemicals Lawesson's Reagent, 97%
Description
Lawesson's reagent is a thiation agent used to convert carbonyl compounds into thiocarbonyls. It is also used to thionate enones, esters, lactones, amides, lactams and quinones. Further, it is used to prepare thiols from alcohols. It is associated with silver perchlorate and utilized as and oxophilic Lewis acid catalyst for Diels-Alder reaction of dienes with alfa, beta-unsaturated aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lawesson′s reagent is a thiation agent used to convert carbonyl compounds into thiocarbonyls. It is also used to thionate enones, esters, lactones, amides, lactams and quinones. Further, it is used to prepare thiols from alcohols. It is associated with silver perchlorate and utilized as and oxophilic Lewis acid catalyst for Diels-Alder reaction of dienes with alfa, beta-unsaturated aldehydes.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 221°C to 225°C |
Quantity | 5 g |
UN Number | UN3134 |
Beilstein | 1024888 |
Sensitivity | Moisture sensitive |
Merck Index | 14,5391 |
Solubility Information | Insoluble in water. |
Formula Weight | 404.47 |
Percent Purity | 97% |
Chemical Name or Material | Lawesson's Reagent |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10238993 | Certificate of Analysis | 07/12/2023 | A14530.14,A14530.36,A14530.22,A14530.06 |
10233049 | Certificate of Analysis | 11/07/2023 | A14530.14,A14530.36,A14530.22,A14530.06,A14530 |
GBFA019096 | Certificate of Analysis | 22/11/2021 | A14530.14,A14530.36,A14530.22,A14530.06 |
10179911 | Certificate of Analysis | 22/11/2021 | A14530.14,A14530.36,A14530.22,A14530.06 |
10220993 | Certificate of Analysis | 22/11/2021 | A14530.14,A14530.36,A14530.22,A14530.06 |
Safety and Handling

- Lawesson's Reagent
- Danger
- Substance which\, in contact with water\, emits flammable gases Category 2
- H261-In contact with water releases flammable gases.
- EUH029-Contact with water liberates toxic gas.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : 2,4-bis(4-methoxyphenyl)-1,3,2,4-dithiadiphosphetane-2,4-disulfide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.