missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
CAS | 19172-47-5 |
---|---|
Molecular Formula | C14H14O2P2S4 |
Molecular Weight (g/mol) | 404.452 |
InChI Key | CFHGBZLNZZVTAY-UHFFFAOYSA-N |
Synonym | lawesson's reagent, lawesson reagent, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane 2,4-disulfide, unii-a4125mq8rx, 1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-, 2,4-disulfide, 2,4-bis 4-methoxyphenyl-2,4-dithioxo-1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane-2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulphide, 4-methoxyphenylthiophosphoric cyclic di thioanhydride |
PubChem CID | 87949 |
IUPAC Name | 2,4-bis(4-methoxyphenyl)-2,4-bis(sulfanylidene)-1,3,2$l^{5},4$l^{5}-dithiadiphosphetane |
SMILES | COC1=CC=C(C=C1)P2(=S)SP(=S)(S2)C3=CC=C(C=C3)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10582611
|
Thermo Scientific Acros
210890250 |
25 g | Glass bottle |
£46.23
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10124840
|
Thermo Scientific Acros
210891000 |
100 g | Glass bottle |
£125.63
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11328901
|
Thermo Scientific Acros
210895000 |
500 g | Glass Bottle |
£433.16
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-S bond formationChemical Identifiers
19172-47-5 | |
404.452 | |
lawesson's reagent, lawesson reagent, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane 2,4-disulfide, unii-a4125mq8rx, 1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-, 2,4-disulfide, 2,4-bis 4-methoxyphenyl-2,4-dithioxo-1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane-2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulphide, 4-methoxyphenylthiophosphoric cyclic di thioanhydride | |
2,4-bis(4-methoxyphenyl)-2,4-bis(sulfanylidene)-1,3,2$l^{5},4$l^{5}-dithiadiphosphetane |
C14H14O2P2S4 | |
CFHGBZLNZZVTAY-UHFFFAOYSA-N | |
87949 | |
COC1=CC=C(C=C1)P2(=S)SP(=S)(S2)C3=CC=C(C=C3)OC |
Specifications
White | |
228.0°C to 231.0°C | |
C14H14O2P2S4 | |
lawesson's reagent, lawesson reagent, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane 2,4-disulfide, unii-a4125mq8rx, 1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-, 2,4-disulfide, 2,4-bis 4-methoxyphenyl-2,4-dithioxo-1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane-2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulphide, 4-methoxyphenylthiophosphoric cyclic di thioanhydride | |
CFHGBZLNZZVTAY-UHFFFAOYSA-N | |
2,4-bis(4-methoxyphenyl)-2,4-bis(sulfanylidene)-1,3,2$l^{5},4$l^{5}-dithiadiphosphetane | |
87949 | |
99% | |
Lawesson's Reagent |
19172-47-5 | |
99% | |
10,39; 12,59; 13,38; 15,37; 16,37 | |
Solubility in water: decomposes. | |
COC1=CC=C(C=C1)P2(=S)SP(=S)(S2)C3=CC=C(C=C3)OC | |
404.452 | |
404.45 | |
Powder |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Lawesson''s Reagent
Signal Word
- Danger
Hazard Category
- Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
- H261-In contact with water releases flammable gases.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P231+P232-Handle under inert gas. Protect from moisture.
- P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
- P314-Get medical advice/attention if you feel unwell.
RUO – Research Use Only