missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ LB Broth (1X)
GREENER_CHOICE

LB broth (Lennox formulation) is a ready-to-use liquid growth medium used for bacterial culture.

Brand:  Gibco™ 10855021

18.49 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11508646

  • £24.33 / 500mL
Estimated Shipment: 03-12-2024
to see stock.
Edge
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

LB broth (Lennox formulation) is a ready-to-use liquid growth medium used for bacterial culture. Gibco LB Broth is filter-sterilized and can be used for nearly all applications that require a liquid bacteria culturing step, such as protein expression, cloning, and expansion.

LB broth is one of the most referenced bacterial media for cultivating and maintaining E. coli strains. Our LB broth is based on the Lennox formulation, one of the most commonly used formulations for LB medium.

LB broth formulation
Formulation per liter: 10 g SELECT Peptone 140, 5 g SELECT Yeast Extract, 5 g sodium chloride.
• Tryptone provides nitrogen, vitamins, minerals, and amino acids essential for growth
• Yeast extract is a source of vitamins, particularly B-group vitamins
• Sodium chloride supplies essential electrolytes for transport and osmotic balance

Primary applications
• Growth and maintenance of E. coli strains for plasmid purification and/or protein expression
• Resuspension of T. pyriformis cell fractions

Gibco media bottles
Each media bottle features an angled opening for simplified pipetting. The 1-L Gibco media bottle uses 39% less source material (33% less for the 500-mL bottle) than comparable media bottles on the market. The cap and bottle are made from high-density polyethylene (HDPE) and polyethylene terephthalate (PETE) and can both be recycled.

Find the media format that you need
We offer other ready-to-use media formulations, such as Terrific Broth (Cat. No. A1374301) or S.O.C Recovery Medium (Cat. No. 15544034).

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Escherichia coli
Bottle
Ready-to-Use
Sterile
Sustainable packaging
Room Temperature
• 500 mL LB Broth (1X)

Store at room temperature.

LB Broth (Lennox)
Liquid Medium
Coliforms
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.