Learn More
Macherey-Nagel™ TLC Spray Visualization Reagents
Used for postchromatographic detection reactions in thin-layer chromatography. Macherey-Nagel™ TLC Spray Visualization Reagents are a selection of frequently used spray reagents that are suited for spraying or dipping TLC plates. Glass laboratory sprayer (mfr. no. 814101) sold separately.
Brand: Macherey-Nagel™ 814402
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Broad selection of premium reagents available for detection of a wide range of substances
- Can be used for spraying or dipping of TLC plates
- Packaged in 100mL screw-top amber bottles
- Use with chemically-resistant sprayers, including Macherey-Nagel glass laboratory sprayer with rubber bulb (mfr. no. 814101, sold separately)
Specifications
7705-08-0 | |
Spraying or Dipping | |
Dragendorff-Munier reagent [Bismuth(III) Nitrate–Potassium Iodide–L(+)-Tartaric Acid] in Water | |
TLC plates | |
Odorless | |
2-3 | |
100 mL |
Colorless | |
Dragendorff-Munier spray reagent | |
Detection of alkaloids and other nitrogen compounds | |
Water | |
Amber bottle with screw cap | |
Spray Visualization Reagent | |
Solution |
Safety and Handling
- spray reagent a.dragendorff-munier
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Supplemental information
- MIXTURE LIST-Contains : bismuth(III) nitrate, basic, potassium iodide,L(+)tartaric acid