missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ TLC Spray Visualization Reagents

Used for postchromatographic detection reactions in thin-layer chromatography. Macherey-Nagel™ TLC Spray Visualization Reagents are a selection of frequently used spray reagents that are suited for spraying or dipping TLC plates. Glass laboratory sprayer (mfr. no. 814101) sold separately.

Brand:  Macherey-Nagel™ 814923


 View more versions of this product

Product Code. 10206521

  • £35.30 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Broad selection of premium reagents available for detection of a wide range of substances
  • Can be used for spraying or dipping of TLC plates
  • Packaged in 100mL screw-top amber bottles
  • Use with chemically-resistant sprayers, including Macherey-Nagel glass laboratory sprayer with rubber bulb (mfr. no. 814101, sold separately)
Specifications

Specifications

7705-08-0
0.79–0.86g/cm3
Rhodamin B spray reagent
Rhodamine B in Ethanol
TLC plates
-114°C
Alcoholic
7
100 mL
Clear
Spraying or Dipping
<1.00% Rhodamin B
Detection of lipids
Ethanol
C28H31ClN2O3
Amber bottle with screw cap
Spray Visualization Reagent
Solution
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Rhodamine B spray reagent

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.

Supplemental information
  • MIXTURE LIST-Contains : ethanol (denatured with 1%IPA/1%MEK, acc.2016/1867/EU)

SDS
Documents

Documents

Certificates
Special Offers

Special Offers