Learn More
Macherey-Nagel™ TLC Spray Visualization Reagents
Used for postchromatographic detection reactions in thin-layer chromatography. Macherey-Nagel™ TLC Spray Visualization Reagents are a selection of frequently used spray reagents that are suited for spraying or dipping TLC plates. Glass laboratory sprayer (mfr. no. 814101) sold separately.
Brand: Macherey-Nagel™ 814206
Code : NEW
Additional Details : Weight : 0.07900kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Description
- Broad selection of premium reagents available for detection of a wide range of substances
- Can be used for spraying or dipping of TLC plates
- Packaged in 100mL screw-top amber bottles
- Use with chemically-resistant sprayers, including Macherey-Nagel glass laboratory sprayer with rubber bulb (mfr. no. 814101, sold separately)
Specifications
7705-08-0 | |
0.79–0.86g/cm3 | |
Rubeanic acid spray reagent | |
Rubeanic Acid in Ethanol | |
TLC plates | |
-114°C | |
Alcoholic | |
7 | |
100 mL |
Yellow | |
Spraying or Dipping | |
0.1 to <1% Rubeanic Acid | |
Detection of heavy metal cations | |
Ethanol | |
C2H4N2S2 | |
Amber bottle with screw cap | |
Spray Visualization Reagent | |
Solution |