missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ TLC Spray Visualization Reagents

Used for postchromatographic detection reactions in thin-layer chromatography. Macherey-Nagel™ TLC Spray Visualization Reagents are a selection of frequently used spray reagents that are suited for spraying or dipping TLC plates. Glass laboratory sprayer (mfr. no. 814101) sold separately.

Brand:  Macherey-Nagel™ 814920


 View more versions of this product

Product Code. 10565371

  • £35.30 / 100mL
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Broad selection of premium reagents available for detection of a wide range of substances
  • Can be used for spraying or dipping of TLC plates
  • Packaged in 100mL screw-top amber bottles
  • Use with chemically-resistant sprayers, including Macherey-Nagel glass laboratory sprayer with rubber bulb (mfr. no. 814101, sold separately)
Specifications

Specifications

7705-08-0
0.785g/cm3
Bromocresol green spray reagent
Bromocresol Green in 2-Propanol
TLC plates
-90°C
Alcoholic
Spray Visualization Reagent
Solution
Green
Spraying or Dipping
0.1 to <1% Bromocresol Green
Detection of organic acids
2-Propanol
C21H14Br4O5S
Amber bottle with screw cap
100 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Bromocresol green spray reagent 100 mL

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.

Supplemental information
  • MIXTURE LIST-Contains : 2-propanol, promocresol green

SDS
Documents

Documents

Certificates
Special Offers

Special Offers