missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ TLC Spray Visualization Reagents

Used for postchromatographic detection reactions in thin-layer chromatography. Macherey-Nagel™ TLC Spray Visualization Reagents are a selection of frequently used spray reagents that are suited for spraying or dipping TLC plates. Glass laboratory sprayer (mfr. no. 814101) sold separately.

Brand:  Macherey-Nagel™ 814302


 View more versions of this product

Product Code. 10609581

  • £35.10 / 100mL
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Broad selection of premium reagents available for detection of a wide range of substances
  • Can be used for spraying or dipping of TLC plates
  • Packaged in 100mL screw-top amber bottles
  • Use with chemically-resistant sprayers, including Macherey-Nagel glass laboratory sprayer with rubber bulb (mfr. no. 814101, sold separately)
Specifications

Specifications

7705-08-0
0.79–0.86g/cm3
Molydatophosphoric acid spray reagent
Molydatophosphoric acid in Ethanol
TLC plates
-114°C
Alcoholic
7
100 mL
Colorless
Spraying or Dipping
3 to <5% Molydatophosphoric Acid
Detection of lipids, sterols, steroids, reducing compounds
Ethanol
H3[P(Mo3O10)4] × H2O
Amber bottle with screw cap
Spray Visualization Reagent
Solution
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • molybdatophosphoric acid spray

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : molybdatophosphoricacid, ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers