missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ TLC Spray Visualization Reagents

Used for postchromatographic detection reactions in thin-layer chromatography. Macherey-Nagel™ TLC Spray Visualization Reagents are a selection of frequently used spray reagents that are suited for spraying or dipping TLC plates. Glass laboratory sprayer (mfr. no. 814101) sold separately.

Brand:  Macherey-Nagel™ 814203

37.30 GBP valid until 2024-12-31
Use promo code "23260" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.08600kg Transport : UN number : 1993 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 10669191

  • £40.55 / 100mL
Estimated Shipment: 09-09-2024
to see stock.
Add to basket

Description

Description

  • Broad selection of premium reagents available for detection of a wide range of substances
  • Can be used for spraying or dipping of TLC plates
  • Packaged in 100mL screw-top amber bottles
  • Use with chemically-resistant sprayers, including Macherey-Nagel glass laboratory sprayer with rubber bulb (mfr. no. 814101, sold separately)
Specifications

Specifications

7705-08-0
0.79–0.86g/cm3
Ninhydrin spray reagent
Ninhydrin in Ethanol
TLC plates
-114°C
Alcoholic
Spray Visualization Reagent
Solution
Red
Spraying or Dipping
0.1 to <1% Ninhydrin
Detection of amino acids, amines, amino sugars
Ethanol
C9H6O4
Amber bottle with screw cap
100 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ninhydrin spray reagent 100 ml

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.

Supplemental information
  • MIXTURE LIST-Contains : ethanol, ninhydrin

SDS
Documents

Documents

Certificates
Special Offers

Special Offers