Learn More
Macherey-Nagel™ TLC Spray Visualization Reagents
Used for postchromatographic detection reactions in thin-layer chromatography. Macherey-Nagel™ TLC Spray Visualization Reagents are a selection of frequently used spray reagents that are suited for spraying or dipping TLC plates. Glass laboratory sprayer (mfr. no. 814101) sold separately.
Brand: Macherey-Nagel™ 814921
Code : NEW
Additional Details : CAS Number : 67-63-0 Weight : 0.07900kg Transport : UN number : 1219 Chem class : 3 Pack group : II
Description
- Broad selection of premium reagents available for detection of a wide range of substances
- Can be used for spraying or dipping of TLC plates
- Packaged in 100mL screw-top amber bottles
- Use with chemically-resistant sprayers, including Macherey-Nagel glass laboratory sprayer with rubber bulb (mfr. no. 814101, sold separately)
Specifications
7705-08-0 | |
0.785g/cm3 | |
2'7'-Dichlorofluorescein spray reagent | |
2',7'-Dichlorofluorescein in 2-Propanol | |
TLC plates | |
-90°C | |
Alcoholic | |
Spray Visualization Reagent | |
Solution |
Green | |
Spraying or Dipping | |
<1.00% 2'7'-Dichlorofluorescein | |
Detection of lipids (saturated, unsaturated) | |
2-Propanol | |
C20H10Cl2O5 | |
Amber bottle with screw cap | |
100 mL |
Safety and Handling
- X1 2',7'-Dichlorofluorescein spr. 100 mL
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Supplemental information
- MIXTURE LIST-Contains : 2-propanol, 2',7'-dichlorfluorescein