missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3 Antibody Pack, Novus Biologicals™
For Research Applications
Brand: Bio-Techne NBP2-59703
Additional Details : Weight : 0.58200kg
Description
Includes: NB100-2220SS: LC3B Antibody, NBP2-46892SS: LC3B Antibody (1251A)Specifications
LC3 | |
1.0mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated | |
ATG8F, LC3B, MAP1A/1BLC3, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta | |
For NB100-2220SS:, Antibody was made to a synthetic peptide made to an N-terminal portion of the human LC3B protein sequence (between residues 1-100). [UniProt# Q9GZQ8].,, For NBP2-46892SS:, Antibody (1251A) was made to a synthetic peptide made to an N-terminal portion of the human LC3B protein sequence (between residues 1-100). [UniProt# Q9GZQ8]. | |
Autophagy, Cancer, Cardiovascular Biology, mTOR Pathway | |
Human, Mouse, Fish, Rabbit, Hamster, Avian, Primate, Bat, Golden Syrian Hamster, Canine, Zebrafish, Mouse, Rat, Rabbit, Alligator, Avian, Bacteria, Bat, Bovine, Canine |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay | |
Unconjugated | |
PBS with | |
MAP1LC3B | |
2 Vials | |
81631 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |