missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | LC3 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Flow Cytometry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16863314
|
Proteintech
14600-1-AP-20UL |
20 μL |
£130.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16853314
|
Proteintech
14600-1-AP-150UL |
150 μL |
£339.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody can cross react with MAP1LC3A, MAP1LC3B, and MAP1LC3C.
LC3B (Autophagy Marker Light Chain 3B) in humans, is encoded by the gene MAP1LC3B (Microtubule-associated proteins 1A/1B light chain 3B). The product of this gene is associated with microtubule assembly and important is in neurogenesis. Recent studies indicates that LC3B plays a vital role in autophagy, a process that involves the bulk degradation of cytoplasmic component. Three human LC3 isoforms undergo post-translational modifications during autophagy.Specifications
LC3 | |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q62625, Q6XVN8, Q91VR7, Q9CQV6, Q9GZQ8, Q9H492 | |
362245, 64862, 66734, 67443, 81631, 84557 | |
LC3 Fusion Protein Ag6144 | |
Primary | |
-20°C | |
MAP1LC3A |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
LC3, LC3B, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, MAP1A/MAP1B light chain 3 B, MAP1ALC3, MAP1LC3B | |
Map1lc3a, Map1lc3b | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |