Learn More
Hach™ LCK Cuvette Tests
Insert, read and finish. Hach™ LCK Cuvette Tests are pre-dosed reagents packaged in a fully equipped box. They provide maximum safety, ease of use and photometric evaluation, while using a minimum amount of reagent/chemicals for an environmentally friendly solution.
Brand: Hach™ LCK362
Description
Hach LCK Cuvette Tests are officially approved for legally required consent limits. With the help of Addista standard solutions and round robin test solutions, they provide the assurance that analytical results are always correct. The photometer identifies the cuvette test from its barcode (IBR+) and calculates the result immediately. Cuvettes provide precision and safety using the Dosicap Zip that eliminated contact with reagents.
- Pre-dosed, ready-to-use reagents with no rinsing steps needed
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Specifications
Acid capacity (KS4.3 / CaCO3) | |
Pre-dosed Tubes | |
Photometer (DR3900, DR6000, DR1900, DR2800, DR3800, DR5000), Addista standard solutions | |
25 | |
15°C to 25°C | |
LCK Cuvette Test |
Blue | |
Water quality testing | |
Fully equipped box, reagent set | |
0.5 to 8.0mmol/L KS4.3 / 25 to 400mg/L CaCO3 | |
Hach Method |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.