missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LCTL Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547801
This item is not returnable.
View return policy
Description
In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse Lactase-like Protein/LCTL is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
LCTL Polyclonal antibody specifically detects LCTL in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
LCTL | |
Polyclonal | |
Unconjugated | |
LCTL | |
KL lactase phlorizin hydrolase; KLG; klotho gamma; klotho/lactase-phlorizin hydrolase-related protein; KLPH; lactase like; lactase-like; lactase-like protein; LCTL; UNQ3022/PRO9820 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
197021 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q6UWM7 | |
LCTL | |
CHO-derived recombinant human Lactase-like Protein/LCTL Arg23-Glu541 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |