missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kautex™ HDPE, Adapter
Click to view available options
For Use With (Equipment):
For Use with 5 to 60 L Carboys
Description
- Use to attach stopcock to Carboy
Specifications
Color | Blue |
For Use With (Equipment) | For Use with 5 to 60 L Carboys |
Material | HDPE |
No. per Pack | 1 Ea. |
Product Type | Adapter |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'