missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kautex™ 500 mL Safety Wash Bottles Series 303 LDPE
Designed to give an accurate fine controlled stream of liquid. Kautex™ 500mL Safety Wash Bottles provide excellent chemical resistance for laboratory safety.
£5.75 - £556.08
Specifications
Capacity (Metric) | 500 mL |
---|---|
Color | Natural |
Diameter (Metric) Inner Neck | 39.2 mm |
Material | LDPE |
No. per Pack | 10 |
Product Code | Brand | Chemical Label | Closure Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Chemical Label | Closure Color | Price | Quantity & Availability | ||||
15708887
|
Kautex™
2000770025 |
Acetone | Red |
£74.65
Pack of 10 |
In Stock
Log in to see stock. |
||||
15768877
|
Kautex™
2000770021 |
Ethanol | Green |
£55.61
Pack of 10 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
||||
15778877
|
Kautex™
2000770022 |
Isopropanol | Yellow |
£83.28
Pack of 10 |
In Stock
Log in to see stock. |
||||
17898097
|
Kautex™
2000770023 |
Methanol | Orange |
£556.08
Pack of 100 |
Estimated Shipment: 10-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15798877
|
Kautex™
2000770024 |
Distilled Water | Blue |
£67.70
Pack of 10 |
In Stock
Log in to see stock. |
||||
15788877
|
Kautex™
2000770023 |
Methanol | Orange |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10146150
|
Kautex™
2000770023 |
Methanol | Orange |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10667691
|
Kautex™
2000770025 |
Acetone | Red |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10248290
|
Kautex™
2000770021 |
Ethanol | Green |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Imprinted with hazard symbols and substances of content in German, English and French language
- Writing space
- Abrasion-resistant printing
- Closure color matches color of ink for easy identification of media
- Leak-proof
- Completely emptied by sliding delivery tube, even when inverted.
Specifications
500 mL | |
39.2 mm | |
10 | |
Wide | |
154 mm | |
77 mm |
Natural | |
LDPE | |
100 Pk. | |
Round | |
Wash Bottle | |
Screw Cap with Delivery Nozzle |