missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ LDPE Round Container
Container for various applications. Produced seamlessly and stress-relieved with the LaboPlast™ sintering method. The wall thickness increases downwardly. This guarantees optimum ratio of wall thickness to filling load. The containers are also color
£446.00
Specifications
Color | Transparent |
---|---|
Material | LDPE |
Shape | Round |
Product Type | Storage Container |
Closure Liner | LDPE |
Description
- L-LDPE (linear LDPE), transparent
- Rupture-proof
Specifications
Transparent | |
Round | |
LDPE |
LDPE | |
Storage Container |