Learn More
Kautex™ LDPE Screw Closure
Feature LDPE light weight, tough construction. Kautex™ LDPE Screw Closures are available in a wide variety of sizes for Kautex™ narrow and wide neck bottles.
Brand:  Kautex™ 2000070608
Additional Details : Weight : 0.60300kg
 View more versions of this product
Product Code. 15991401
  • £42.55 / Pack of 100
In Stock
to see stock.
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Made from low density polyethylene
  • Translucent
  • Available for a wide variety of sizes
Specifications
Specifications
40 mm
200/250/300 mL Wide Mouth Bottle
Natural
LDPE
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products