missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ LDPE Wash Bottles

Product Code. 11705318
Click to view available options
Capacity:
500mL
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

OSHA-compliant, labeled safety wash bottles with a built-in, nonmetallic vent to release pressure and prevent leaking. Cole-Parmer™ LDPE Wash Bottles feature chemically resistant silk-screened labels.

  • Bottles display name, chemical formula, NFPS hazard rating, and DOT symbols for primary health hazard of contents
  • View contents through translucent bottle
  • Easily identify contents by color-coded closures

Specifications

Capacity 500mL
Color Translucent
Includes One of each-Acetone, Ethanol, Isopropanol, Methanol
Material LDPE
No. per Pack 5/Pk
Autoclavable Not Autoclavable
Height (Metric) 21.3 cm
Product Type Wash Bottle
Closure Material Polypropylene
Closure Size 28 mm
Diameter (Metric) 7 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'