missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Le Mans S1P Shoe

Product Code. 17502486
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 17502486

Brand: Emma Safety Footwear MM70354740

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Like its name suggests, you could wear the Le Mans for 24 hr. as the shoes offer optimal ventilation, perspiration and moisture removal with the Hydro-Tec™ antibacterial lining material.

  • Metal Free
  • Dry, fresh feet for up to 24 hr.The name already suggests it: you can safely wear LeMans for 24 hours.
  • The shoe offers optimal ventilation and moisture is immediately removed by the Hydro-Tec™ antibacterial lining.
  • LeMans is ideal for indoor working environments and work in transport and logistics.
  • LeMans looks sporty, that's how the shoe feels; soles with energy return and a fit that hugs the feet perfectly.
  • Industries:
    • Logistics & Transport
    • Electronics
    • Light Industry
  • Safety shoe
    • S1P-Safety shoe for dry conditions

Especificaciones

Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Black, Gray, Orange
For Use With (Application) Logistics and Transport, Electronics, Manufacturing
Liner Material Hydro-Tec Antibacterial
Size 40
Fit Standard Width
Ankle Style Low-cut
Product Type Safety Shoe
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado