missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Le Mans S1P Shoe

Product Code. 17532486
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 17532486

Brand: Emma Safety Footwear MM70354743

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Like its name suggests, you could wear the Le Mans for 24 hr. as the shoes offer optimal ventilation, perspiration and moisture removal with the Hydro-Tec™ antibacterial lining material.

  • Metal Free
  • Dry, fresh feet for up to 24 hr.The name already suggests it: you can safely wear LeMans for 24 hours.
  • The shoe offers optimal ventilation and moisture is immediately removed by the Hydro-Tec™ antibacterial lining.
  • LeMans is ideal for indoor working environments and work in transport and logistics.
  • LeMans looks sporty, that's how the shoe feels; soles with energy return and a fit that hugs the feet perfectly.
  • Industries:
    • Logistics & Transport
    • Electronics
    • Light Industry
  • Safety shoe
    • S1P-Safety shoe for dry conditions

Specifications

Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Black, Gray, Orange
For Use With (Application) Logistics and Transport, Electronics, Manufacturing
Liner Material Hydro-Tec Antibacterial
Size 43
Fit Standard Width
Ankle Style Low-cut
Product Type Safety Shoe
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.