missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead foil, 0.76mm (0.03in) thick, 99.8% (metals basis)
Click to view available options
Quantity:
15 x 15 cm
30 x 30 cm
Unit Size:
Each
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Gray |
| Appearance | Gray foil |
| Quantity | 30 x 30 cm |
| Odor | Odorless |
| Assay Percent Range | 99.8% (Metals basis) |
| Form | Foil |
| Chemical Name or Material | Lead |
Safety and Handling
Product Identifier
- Lead foil
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- Reproductive toxicity on or via lactation
- Reproductive toxicity Category 1A
- Specific target organ toxicity Category 1
- H360FD-May damage fertility. May damage the unborn child.
- H362-May cause harm to breast-fed children.
- H372-Causes damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction