missing translation for 'onlineSavingsMsg'
Learn More

3M™ Lead Free Steam Indicator Tape for Disposable Wraps

Product Code. 12916986
Click to view available options
Dimensions (L x W):
55 m x 18 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12916986

Brand: 3M™ 7000138233

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provides an exposure monitor that securely seals packs and provides a visual assurance that the package has been exposed to the steam sterilization process. 3M™ Lead Free Steam Indicator Tape for Disposable Wraps utilizes a trusted, high quality adhesive.

  • Lead free ink means no hazardous waste issues
  • Stretchable backing that minimizes tape “pop off” during sterilization
  • Not made with natural rubber latex

Specifications

Dimensions (L x W) 55 m x 18 mm
Length (Metric) 55 m
Product Type Indicator Tape
Quantity 1 Pk.
Width (Metric) Exterior 18 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.