missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead granules, 99.5% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Gray |
Appearance | Gray granules |
Assay Percent Range | 99.5% min |
Quantity | 100 g |
Odor | Odorless |
Form | Granular |
Chemical Name or Material | Lead |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N24L001 | Certificate of Analysis | 26/02/2025 | 036227.36,036227.A3,036227.22 |
U29J023 | Certificate of Analysis | 21/09/2023 | 036227.36,036227.A3,036227.22 |
U19F059 | Certificate of Analysis | 21/06/2022 | 036227.36,036227.A3,036227.22,036244.03,036244.04 |
P29I051 | Certificate of Analysis | 12/04/2022 | 036227.36,036227.A3,036227.22,036244.03,036244.04 |
G13K31 | Certificate of Analysis | 18/11/2021 | 036227.36,036227.A3,036227.22,036244.03,036244.04 |
Safety and Handling



Product Identifier
- Lead granules
- Danger
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lead granules, 99.5% min, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction