Learn More
Lead(II) 2-ethylhexanoate, Thermo Scientific Chemicals
Description
Films of lead (II) 2-ethylhexanote can be cast on silicon solutions by spin coating. Lead (II) 2-ethylhexanote is a common solvent in metal-organic deposition (MOD) process. Intermetallic PtPb nanoparticles have been synthesized by the chemical reduction of dimethyl(1,5-cyclooctadiene)platinum and lead(II)2-ethylhexanoate by sodium naphthalide in THF or diglyme. The precursors used for the photochemical production of PZT or PLZT films were lead(II) 2-ethylhexanoate, lanthanum(III) 2-ethylhexanoate, zirconyl(IV) 2-ethylhexa- noate, and titanium(IV) isopropoxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.56 g/mL |
Odor | Odorless |
Quantity | 50 g |
UN Number | UN3082 |
Solubility Information | Insoluble in water. |
Formula Weight | 493.61 |
Physical Form | Viscous Liquid |
Chemical Name or Material | Lead(II) 2-ethylhexanoate |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS09_Aquatic_Toxicity.png)
- Lead(II) 2-ethylhexanoate
- Danger
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P501-Dispose of contents/container to .
RUO – Research Use Only