missing translation for 'onlineSavingsMsg'
Learn More
Lead(II) Acetate Trihydrate, Certified AR for Analysis, Fisher Chemical™ Product Code.: 10407840

Lead(II) Acetate Trihydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10407840
250 g
Click to view available options
Quantity:
250 g
500 g
This item is not returnable. View return policy

Product Code. 10407840

Brand: Fisher Chemical L/1000/50

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 75°C
Color White
pH 5.5 to 6.5
Packaging Plastic powder jar
Quantity 250 g
IUPAC Name λ²-lead(2+) diacetate trihydrate
Formula Weight 379.33g/mol
Physical Form Solid
compliance-icons compliance-icons
Product Identifier
  • Lead(II) acetate trihydrate
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH201-Contains lead. Should not be used on surfaces liable to be chewed or sucked by children.
Precautionary Statement
  • P281-Use personal protective equipment as required.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(II) Acetate Trihydrate, Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.