Learn More
Lead(II) acetate trihydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
£19.60 - £350.75
Chemical Identifiers
CAS | 6080-56-4 |
---|---|
Molecular Formula | C4H12O7Pb |
Molecular Weight (g/mol) | 379.30 |
MDL Number | MFCD00150023 |
InChI Key | MCEUZMYFCCOOQO-UHFFFAOYSA-L |
Synonym | lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate |
PubChem CID | 22456 |
ChEBI | CHEBI:33112 |
IUPAC Name | lead(2+);diacetate;trihydrate |
SMILES | O.O.O.[Pb++].CC([O-])=O.CC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11356787
|
Thermo Scientific Alfa Aesar
010719.06 |
5 g |
£19.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376787
|
Thermo Scientific Alfa Aesar
010719.18 |
50 g |
£73.57
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11366787
|
Thermo Scientific Alfa Aesar
010719.30 |
250 g |
£350.75
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lead(II) acetate trihydrate is used as a mordant in textile printing and dyeing, drier in paints, varnishes and as a water repellant. It is also used in cosmetics and to prepare other lead compounds. It serves as an ingredient in progressive types of hair coloring dyes. Further, it is used in the detection of poisonous gas hydrogen sulfide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6080-56-4 | |
379.30 | |
MCEUZMYFCCOOQO-UHFFFAOYSA-L | |
22456 | |
lead(2+);diacetate;trihydrate |
C4H12O7Pb | |
MFCD00150023 | |
lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate | |
CHEBI:33112 | |
O.O.O.[Pb++].CC([O-])=O.CC([O-])=O |
Specifications
75°C | |
Crystalline | |
MFCD00150023 | |
3730298 | |
lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate | |
MCEUZMYFCCOOQO-UHFFFAOYSA-L | |
lead(2+);diacetate;trihydrate | |
22456 | |
379.33 (325.28 Anhydrous) | |
Puratronic™ | |
(metals basis) |
6080-56-4 | |
C4H12O7Pb | |
UN1616 | |
14,5397 | |
Freely soluble in water,alcohol and glycerol. | |
O.O.O.[Pb++].CC([O-])=O.CC([O-])=O | |
379.30 | |
CHEBI:33112 | |
99.995% | |
2.55 g/mL | |
Lead(II) acetate trihydrate |
Safety and Handling
GHS H Statement
H360-H373
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H360D-H361f-H372
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD ACETATE
missing translation for 'einecsNumber' : 206-104-4
missing translation for 'rtecsNumber' : OF8050000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Lead(II) acetate trihydrate, Puratronic
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only