Learn More
Lead(II) bromide, 98+%, Thermo Scientific Chemicals
£26.43 - £66.06
Chemical Identifiers
CAS | 10031-22-8 |
---|---|
Molecular Formula | Br2Pb |
Molecular Weight (g/mol) | 367.00 |
MDL Number | MFCD00011156 |
InChI Key | ZASWJUOMEGBQCQ-UHFFFAOYSA-L |
Synonym | lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2xShow More |
PubChem CID | 24831 |
SMILES | [Br-].[Br-].[Pb++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11424007
|
Thermo Scientific Alfa Aesar
A19406.22 |
100 g |
£26.43
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11434007
|
Thermo Scientific Alfa Aesar
A19406.36 |
500 g |
£66.06
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It finds its uses as a catalyst to make polyesters, a filler for polypropylene (flame-resistant), and in glass optical waveguides. It also finds ints use in laboratories and metallurgy. it is used as acryl amide monomer light aggregation function of catalysts and can be used for photoelectric material and heat power semiconductor materials; it is also used as analytical reagent, additive and in the preparation of massicot pigment; it is also used as pigment ingredient and used for printing and dyeing industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its uses as a catalyst to make polyesters, a filler for polypropylene (flame-resistant), and in glass optical waveguides. It also finds ints use in laboratories and metallurgy. it is used as acryl amide monomer light aggregation function of catalysts and can be used for photoelectric material and heat power semiconductor materials; it is also used as analytical reagent, additive and in the preparation of massicot pigment; it is also used as pigment ingredient and used for printing and dyeing industry.
Solubility
Soluble in water at 20°C 5g/L. Insoluble in ethanol. Solubility in N,N-DMF is almost transparent.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
10031-22-8 | |
367.00 | |
ZASWJUOMEGBQCQ-UHFFFAOYSA-L | |
24831 |
Br2Pb | |
MFCD00011156 | |
lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2xShow More | |
[Br-].[Br-].[Pb++] |
Specifications
916°C | |
10031-22-8 | |
PbBr2 | |
UN2291 | |
lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x | |
ZASWJUOMEGBQCQ-UHFFFAOYSA-L | |
dibromolead | |
24831 | |
≥98% | |
6.66 |
373°C | |
Br2Pb | |
MFCD00011156 | |
14,5403 | |
Soluble in water at 20°C 5g/L. Insoluble in ethanol. Solubility in N,N-DMF is almost transparent. | |
[Br-].[Br-].[Pb++] | |
367.00 | |
367.01 | |
Odorless | |
Lead(II) bromide |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P312-P314-P330-P501c
H302+H332-H360Df-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
missing translation for 'einecsNumber' : 233-084-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Lead(II) bromide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.