missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) carbonate, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifica
Melting Point | 400.0°C |
Quantity | 500 g |
Linear Formula | PbCO3 |
Merck Index | 10,1006 |
Solubility Information | Solubility in water: insoluble |
IUPAC Name | lead(2+);carbonate |
Formula Weight | 267.2 |
Grade | ACS Reagent |
Packaging | Plastic bottle |
Chemical Name or Material | Lead(II) carbonate |
Sicurezza e movimentazione
Identificativo prodotto
- Lead(II) carbonate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'