missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) carbonate, ACS, Thermo Scientific Chemicals

Product Code. 11319257
Click to view available options
Quantity:
100 g
500 g
4 x 500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Maximum level of impurities: Insoluble in dilute acetic acid 0.02%, Cl 0.002%, Nitrate and nitrite (as NO3) P.T. (limit about 0.005%), Ca 0.01%, K 0.02%, Na 0.05%, Cd 0.002%, Fe 0.005%, Zn 0.003%

Lead(II) carbonate is used in polymerization catalysts. It serves as a curing agent for silicon resins, heat-resistant lubricants, lead-acid storage batteries, wire-reinforced hoses, thermistors and heat stabilizers for polyvinyl chloride polymers. Further, it is used in electrophotography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-63-0
Molecular Formula CO3Pb
Molecular Weight (g/mol) 267.20
MDL Number MFCD00064631
InChI Key MFEVGQHCNVXMER-UHFFFAOYSA-L
Synonym lead carbonate, lead ii carbonate, cerussete, cerussite, lead 2+ carbonate, plumbous carbonate, dibasic lead carbonate, unii-43m0p24l2b, carbonic acid, lead 2+ salt 1:1, lead carbonate pbco3
PubChem CID 11727
SMILES [Pb++].[O-]C([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 100 g
Solubility Information Soluble in acid and alkali. Insoluble in water,alcohol and ammonia.
IUPAC Name lead(2+);carbonate
Formula Weight 267.2
Grade ACS
Odor Odorless
Appearance White powder
Density 6.6g/cm3 at 20°C
Chemical Name or Material Lead(II) carbonate, For analysis ACS
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) carbonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'